Assay Details
| Gene Symbol: LOC719436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124675.2 | XP_014980161.2 | 659 | 74 | ||
| XM_015124676.2 | XP_014980162.2 | 745 | 74 | |||
| XM_015124678.2 | XP_014980164.2 | 499 | 74 | |||
| XM_028839831.1 | XP_028695664.1 | 495 | 74 | |||
More Information
Set Membership: |
|

LOC719436
Genomic Map