Assay Details
| Gene Symbol: MAGEB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127124.2 | XP_014982610.1 | 186 | 69 | ||
| GenBank mRNA | JV061201.1 | - | 76 | 69 | ||
| JV651578.1 | - | 64 | 69 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015127124.2 transcripts.
Set Membership: |
|

MAGEB2
Genomic Map