Assay Details
Gene Symbol: CRPPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001105001.4 | XP_001105001.2 | 1009 | 97 | ||
XM_015133928.2 | XP_014989414.1 | 1009 | 97 | |||
GenBank mRNA | JU325882.1 | - | 734 | 97 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
