Assay Details
| Gene Symbol: HOXA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002803358.3 | XP_002803404.3 | 943 | 93 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOXA13| Gene Symbol: HOXA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002803358.3 | XP_002803404.3 | 943 | 93 | ||
Set Membership: |
|