Assay Details
| Gene Symbol: DUOXA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015142140.2 | XP_014997626.2 | 1013 | 60 | ||
| GenBank mRNA | JV665036.1 | - | 213 | 60 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DUOXA1| Gene Symbol: DUOXA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015142140.2 | XP_014997626.2 | 1013 | 60 | ||
| GenBank mRNA | JV665036.1 | - | 213 | 60 | ||
Set Membership: |
|