Assay Details
Gene Symbol: ZNF461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015123620.2 | XP_014979106.1 | 245 | 104 | ||
XM_015123621.2 | XP_014979107.1 | 245 | 104 | |||
XM_015123622.2 | XP_014979108.1 | 245 | 104 | |||
XM_028839033.1 | XP_028694866.1 | 245 | 104 | |||
GenBank mRNA | JV665563.1 | - | 215 | 104 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015123620.2, XM_015123621.2, XM_015123622.2, XM_028839033.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
