Assay Details
Gene Symbol: ZNF461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015123620.2 | XP_014979106.1 | 463 | 93 | ||
XM_015123622.2 | XP_014979108.1 | 426 | 93 | |||
XM_028839031.1 | XP_028694864.1 | 374 | 93 | |||
XM_028839032.1 | XP_028694865.1 | 337 | 93 | |||
GenBank mRNA | JV665563.1 | - | 433 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
