Assay Details
Gene Symbol: ZNF461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015123620.2 | XP_014979106.1 | 534 | 124 | ||
XM_015123622.2 | XP_014979108.1 | 497 | 124 | |||
XM_028839031.1 | XP_028694864.1 | 445 | 124 | |||
XM_028839032.1 | XP_028694865.1 | 408 | 124 | |||
GenBank mRNA | JV525660.1 | - | 49 | 124 | ||
JV665563.1 | - | 504 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() |
