Assay Details
| Gene Symbol: ATF7IP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125605.2 | XP_014981091.1 | 1128 | 114 | ||
| XM_015125606.2 | XP_014981092.1 | 1128 | 114 | |||
| XM_028841425.1 | XP_028697258.1 | 1036 | 114 | |||
| XM_028841426.1 | XP_028697259.1 | 1128 | 114 | |||
| GenBank mRNA | JV071973.1 | - | 1093 | 114 | ||
| JV662013.1 | - | 1149 | 114 | |||
More Information
Set Membership: |
|

ATF7IP2
Genomic Map