Assay Details
| Gene Symbol: PRRT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125950.2 | XP_014981436.2 | 1178 | 69 | ||
| XM_028840666.1 | XP_028696499.1 | 1155 | 69 | |||
| GenBank mRNA | JU323237.1 | - | 1090 | 69 | ||
| JV564957.1 | - | 1053 | 69 | |||
| JV602459.1 | - | 1135 | 69 | |||
More Information
Set Membership: |
|

PRRT2
Genomic Map