Assay Details
| Gene Symbol: SLC25A38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131305.2 | XP_014986791.1 | 920 | 88 | ||
| XM_015131306.2 | XP_014986792.1 | 920 | 88 | |||
| XM_015131307.2 | XP_014986793.1 | 1561 | 88 | |||
| XM_028843976.1 | XP_028699809.1 | 1034 | 88 | |||
| XM_028843977.1 | XP_028699810.1 | 1034 | 88 | |||
| XM_028843978.1 | XP_028699811.1 | 2172 | 88 | |||
| GenBank mRNA | JU335175.1 | - | 642 | 88 | ||
| JU476166.1 | - | 655 | 88 | |||
| JV047475.1 | - | 511 | 88 | |||
More Information
Set Membership: |
|

SLC25A38
Genomic Map