Assay Details
| Gene Symbol: EIF4EBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145167.2 | XP_015000653.1 | 570 | 76 | ||
| GenBank mRNA | JU474602.1 | - | 340 | 76 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EIF4EBP1| Gene Symbol: EIF4EBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145167.2 | XP_015000653.1 | 570 | 76 | ||
| GenBank mRNA | JU474602.1 | - | 340 | 76 | ||
Set Membership: |
|