Assay Details
| Gene Symbol: KRT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002798598.3 | XP_002798644.2 | 1113 | 75 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT3| Gene Symbol: KRT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002798598.3 | XP_002798644.2 | 1113 | 75 | ||