Assay Details
| Gene Symbol: FSCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015120584.2 | XP_014976070.2 | 2785 | 77 | ||
| GenBank mRNA | JV577478.1 | - | 214 | 77 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FSCN2| Gene Symbol: FSCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015120584.2 | XP_014976070.2 | 2785 | 77 | ||
| GenBank mRNA | JV577478.1 | - | 214 | 77 | ||
Set Membership: |
|