Assay Details
| Gene Symbol: SEPTIN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015120460.2 | XP_014975946.2 | 1519 | 109 | ||
| XM_015120462.2 | XP_014975948.2 | 1243 | 109 | |||
| XM_015120463.2 | XP_014975949.2 | 1681 | 109 | |||
| XM_015120464.2 | XP_014975950.2 | 1903 | 109 | |||
| XM_015120465.2 | XP_014975951.1 | 658 | 109 | |||
| XM_015120466.2 | XP_014975952.1 | 953 | 109 | |||
| XM_015120467.2 | XP_014975953.1 | 762 | 109 | |||
| XM_015120468.2 | XP_014975954.1 | 555 | 109 | |||
| XM_028836849.1 | XP_028692682.1 | 988 | 109 | |||
| XM_028836850.1 | XP_028692683.1 | 493 | 109 | |||
| GenBank mRNA | JU322456.1 | - | 1246 | 109 | ||
| JV045986.1 | - | 1298 | 109 | |||
| JV045987.1 | - | 1303 | 109 | |||
| JV527780.1 | - | 1147 | 109 | |||
More Information
Set Membership: |
|

SEPTIN9
Genomic Map