Assay Details
| Gene Symbol: PCSK1N | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127374.2 | XP_014982860.1 | 715 | 74 | ||
| XM_028841939.1 | XP_028697772.1 | 817 | 74 | |||
| GenBank mRNA | JV578674.1 | - | 557 | 74 | ||
| JV648548.1 | - | 607 | 74 | |||
More Information
Set Membership: |
|

PCSK1N
Genomic Map