Assay Details
| Gene Symbol: SLC25A44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260572.2 | NP_001247501.1 | 766 | 66 | ||
| XM_028846130.1 | XP_028701963.1 | 637 | 66 | |||
| GenBank mRNA | JU337265.1 | - | 776 | 66 | ||
| JU477971.1 | - | 766 | 66 | |||
| JV528725.1 | - | 743 | 66 | |||
| JV599297.1 | - | 777 | 66 | |||
| JV634423.1 | - | 750 | 66 | |||
More Information
Set Membership: |
|

SLC25A44
Genomic Map