Assay Details
| Gene Symbol: IQCF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277943.1 | NP_001264872.1 | 62 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001277943.1 transcripts.
Set Membership: |
|

IQCF5
Genomic Map