Assay Details
| Gene Symbol: PIGP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015132906.2 | XP_014988392.1 | 200 | 88 | ||
| XM_015132907.2 | XP_014988393.1 | 256 | 88 | |||
| XM_015132908.2 | XP_014988394.1 | 229 | 88 | |||
| GenBank mRNA | JV062730.1 | - | 222 | 88 | ||
| JV567750.1 | - | 109 | 88 | |||
More Information
Set Membership: |
|

PIGP
Genomic Map