Assay Details
| Gene Symbol: ZNF366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015140265.2 | XP_014995751.2 | 1713 | 65 | ||
| GenBank mRNA | JU481568.1 | - | 420 | 65 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF366| Gene Symbol: ZNF366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015140265.2 | XP_014995751.2 | 1713 | 65 | ||
| GenBank mRNA | JU481568.1 | - | 420 | 65 | ||
Set Membership: |
|