Assay Details
| Gene Symbol: TOMM20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261051.1 | NP_001247980.1 | 253 | 67 | ||
| GenBank mRNA | JU319662.1 | - | 293 | 67 | ||
| JU473068.1 | - | 350 | 67 | |||
| JV044713.1 | - | 253 | 67 | |||
| JV565490.1 | - | 348 | 67 | |||
More Information
Set Membership: |
|

TOMM20
Genomic Map