Assay Details
| Gene Symbol: RHBDL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015135813.2 | XP_014991299.1 | 684 | 92 | ||
| XM_015135819.2 | XP_014991305.1 | 688 | 92 | |||
| GenBank mRNA | JU335158.1 | - | 562 | 92 | ||
| JU335159.1 | - | 562 | 92 | |||
| JU335160.1 | - | 562 | 92 | |||
More Information
Set Membership: |
|

RHBDL2
Genomic Map