Assay Details
| Gene Symbol: C7H14orf180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144588.2 | XP_015000074.2 | 320 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015144588.2 transcripts.
Set Membership: |
|

C7H14orf180
Genomic Map