Assay Details
| Gene Symbol: PFN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001099519.4 | XP_001099519.1 | 867 | 70 | ||
| XM_015111815.2 | XP_014967301.1 | 711 | 70 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PFN4| Gene Symbol: PFN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001099519.4 | XP_001099519.1 | 867 | 70 | ||
| XM_015111815.2 | XP_014967301.1 | 711 | 70 | |||
Set Membership: |
|