Assay Details
| Gene Symbol: LRRC63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121039.2 | XP_014976525.2 | 563 | 81 | ||
| XM_015121041.2 | XP_014976527.2 | 434 | 81 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRRC63| Gene Symbol: LRRC63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121039.2 | XP_014976525.2 | 563 | 81 | ||
| XM_015121041.2 | XP_014976527.2 | 434 | 81 | |||
Set Membership: |
|