Assay Details
| Gene Symbol: DNAJB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001112237.3 | XP_001112237.2 | 797 | 72 | ||
| XM_015123143.2 | XP_014978629.1 | 1010 | 72 | |||
| XM_015123144.2 | XP_014978630.1 | 823 | 72 | |||
| XM_015123145.2 | XP_014978631.1 | 799 | 72 | |||
| XM_028839660.1 | XP_028695493.1 | 932 | 72 | |||
| GenBank mRNA | JU334302.1 | - | 833 | 72 | ||
| JU475411.1 | - | 840 | 72 | |||
| JV601153.1 | - | 863 | 72 | |||
| JV669354.1 | - | 842 | 72 | |||
More Information
Set Membership: |
|

DNAJB1
Genomic Map