Assay Details
| Gene Symbol: MIR219-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032049.1 | 57 | 68 | |||
| Gene Symbol: RING1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136067.2 | XP_014991553.1 | 52 | 68 | ||
| XM_028846856.1 | XP_028702689.1 | 52 | 68 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome |
|---|
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015136067.2, XM_028846856.1 transcripts.
Set Membership: |
|

MIR219-1
Genomic Map