Assay Details
| Gene Symbol: MIR484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032535.1 | 23 | 55 | |||
| Gene Symbol: NDE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028840593.1 | XP_028696426.1 | 66 | 55 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome |
|---|
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028840593.1 transcripts.
Set Membership: |
|

MIR484
Genomic Map