Assay Details
Gene Symbol: EVI2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015119010.2 | XP_014974496.1 | 384 | 99 | ||
GenBank mRNA | JU340258.1 | - | 220 | 99 | ||
JU482371.1 | - | 191 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015119010.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
