Assay Details
| Gene Symbol: ZNF474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001089304.4 | XP_001089304.3 | 182 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001089304.4 transcripts.
Set Membership: |
|

ZNF474
Genomic Map