Assay Details
| Gene Symbol: H2AJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193866.3 | NP_001180795.1 | 108 | 56 | ||
| GenBank mRNA | JU321876.1 | - | 88 | 56 | ||
| JU472178.1 | - | 94 | 56 | |||
| JV043254.1 | - | 44 | 56 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001193866.3 transcripts.
Set Membership: |
|

H2AJ
Genomic Map