Assay Details
Gene Symbol: HCAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145254.3 | NP_001138726.3 | 922 | 61 | ||
GenBank mRNA | EU809466.1 | - | 922 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
