Assay Details
| Gene Symbol: PPY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193429.3 | NP_001180358.1 | 196 | 78 | ||
| XM_015119756.2 | XP_014975242.1 | 326 | 78 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPY| Gene Symbol: PPY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193429.3 | NP_001180358.1 | 196 | 78 | ||
| XM_015119756.2 | XP_014975242.1 | 326 | 78 | |||
Set Membership: |
|