Assay Details
| Gene Symbol: PAQR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194535.3 | NP_001181464.2 | 1419 | 83 | ||
| XM_028843359.1 | XP_028699192.1 | 1308 | 83 | |||
| GenBank mRNA | JV713858.1 | - | 1043 | 83 | ||
More Information
Set Membership: |
|

PAQR9
Genomic Map