Assay Details
| Gene Symbol: CLDN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194564.3 | NP_001181493.1 | 619 | 81 | ||
| GenBank mRNA | JU470723.1 | - | 381 | 81 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLDN4| Gene Symbol: CLDN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194564.3 | NP_001181493.1 | 619 | 81 | ||
| GenBank mRNA | JU470723.1 | - | 381 | 81 | ||
Set Membership: |
|