Assay Details
| Gene Symbol: RPRML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194690.4 | NP_001181619.1 | 222 | 109 | ||
| GenBank mRNA | JU334450.1 | - | 92 | 109 | ||
| JV667258.1 | - | 226 | 109 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194690.4 transcripts.
Set Membership: |
|

RPRML
Genomic Map