Assay Details
| Gene Symbol: SIGLECL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194706.3 | NP_001181635.2 | 806 | 67 | ||
| XM_015124423.2 | XP_014979909.2 | 757 | 67 | |||
| XM_015124424.2 | XP_014979910.2 | 854 | 67 | |||
| XM_015124426.2 | XP_014979912.2 | 835 | 67 | |||
| XM_015124431.2 | XP_014979917.2 | 475 | 67 | |||
| XM_015124432.2 | XP_014979918.2 | 553 | 67 | |||
| XM_028838533.1 | XP_028694366.1 | 794 | 67 | |||
More Information
Set Membership: |
|

SIGLECL1
Genomic Map