Assay Details
| Gene Symbol: SERPINB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202551.2 | NP_001189480.1 | 52 | 105 | ||
| GenBank mRNA | CO583313.1 | - | 54 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001202551.2 transcripts.
Set Membership: |
|

SERPINB6
Genomic Map