Assay Details
| Gene Symbol: RBIS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257266.1 | NP_001244195.1 | 38 | 65 | ||
| GenBank mRNA | CK231716.1 | - | 64 | |||
| JU326150.1 | - | 1437 | 65 | |||
| JU471005.1 | - | 72 | 65 | |||
| JV045731.1 | - | 92 | 65 | |||
| JV635885.1 | - | 86 | 65 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001257266.1 transcripts.
Set Membership: |
|

RBIS
Genomic Map