Assay Details
| Gene Symbol: GJB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257617.1 | NP_001244546.1 | 139 | 68 | ||
| GenBank mRNA | JU319803.1 | - | 139 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001257617.1 transcripts.
Set Membership: |
|

GJB1
Genomic Map