Assay Details
| Gene Symbol: DDT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257668.1 | NP_001244597.1 | 177 | 78 | ||
| GenBank mRNA | JU332157.1 | - | 177 | 78 | ||
| JU474508.1 | - | 135 | 78 | |||
| JV046319.1 | - | 146 | 78 | |||
More Information
Set Membership: |
|

DDT
Genomic Map