Assay Details
| Gene Symbol: NDUFAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257820.1 | NP_001244749.1 | 187 | 55 | ||
| GenBank mRNA | JU334641.1 | - | 187 | 55 | ||
| JV047147.1 | - | 203 | 55 | |||
| JV507828.1 | - | 86 | 55 | |||
More Information
Set Membership: |
|

NDUFAB1
Genomic Map