Assay Details
| Gene Symbol: COX14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261469.1 | NP_001248398.1 | 194 | 62 | ||
| GenBank mRNA | JU336811.1 | - | 178 | 62 | ||
| JU495231.1 | - | 194 | 62 | |||
| JV044760.1 | - | 194 | 62 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001261469.1 transcripts.
Set Membership: |
|

COX14
Genomic Map