Assay Details
| Gene Symbol: NEUROD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265801.1 | NP_001252730.1 | 43 | 63 | ||
| GenBank mRNA | JV667405.1 | - | 43 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001265801.1 transcripts.
Set Membership: |
|

NEUROD2
Genomic Map