Assay Details
| Gene Symbol: RPS15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265922.1 | NP_001252851.1 | 60 | 80 | ||
| GenBank mRNA | JU491184.1 | - | 71 | 80 | ||
| JV600227.1 | - | 60 | 80 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001265922.1 transcripts.
Set Membership: |
|

RPS15
Genomic Map