Assay Details
| Gene Symbol: RPA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266105.1 | NP_001253034.1 | 221 | 93 | ||
| GenBank mRNA | JU323398.1 | - | 221 | 93 | ||
| JV658099.1 | - | 283 | 93 | |||
More Information
Set Membership: |
|

RPA3
Genomic Map