Assay Details
| Gene Symbol: SLURP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266622.2 | NP_001253551.1 | 68 | 103 | ||
| XM_028852098.1 | XP_028707931.1 | 1065 | 103 | |||
| GenBank mRNA | JV045600.1 | - | 68 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001266622.2, XM_028852098.1 transcripts.
Set Membership: |
|

SLURP2
Genomic Map