Assay Details
| Gene Symbol: SOCS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266928.1 | NP_001253857.1 | 205 | 90 | ||
| GenBank mRNA | JU337260.1 | - | 203 | 90 | ||
| JU477967.1 | - | 215 | 90 | |||
| JV044297.1 | - | 205 | 90 | |||
| JV566825.1 | - | 162 | 90 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001266928.1 transcripts.
Set Membership: |
|

SOCS5
Genomic Map