Assay Details
| Gene Symbol: OPRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321097.2 | NP_001308026.2 | 801 | 76 | ||
| NM_001321098.2 | NP_001308027.2 | 801 | 76 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OPRK1| Gene Symbol: OPRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321097.2 | NP_001308026.2 | 801 | 76 | ||
| NM_001321098.2 | NP_001308027.2 | 801 | 76 | |||
Set Membership: |
|