Assay Details
| Gene Symbol: MT1X | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301313.1 | NP_001288242.1 | 89 | 117 | ||
| GenBank mRNA | CN801994.1 | - | 80 | 117 | ||
| JV582510.1 | - | 89 | 117 | |||
| JV669169.1 | - | 91 | 117 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001301313.1 transcripts.
Set Membership: |
|

MT1X
Genomic Map